Order Entry
Italy
ContactUsLinkComponent
 
Irritant
Methyl-3,4-dichlorobenzoate ≥97%
Catalog # B20027.06
CAS Number:  
Irritant
Methyl-3,4-dichlorobenzoate ≥97%
Catalog # B20027.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:2905-68-2
 :
Formula:C₈H₆Cl₂O₂
MDL Number:MFCD00018165
 :205.04 g/mol
Melting Pt:44…46 °C
 :GHS07
Storage Temperature:Ambient