Order Entry
Italy
ContactUsLinkComponent
Neodymium(III) trifluoromethanesulphonate 98%, REacton®
Irritant
Neodymium(III) trifluoromethanesulphonate 98%, REacton®
Catalog # 43393.09
CAS Number:  
Irritant
Neodymium(III) trifluoromethanesulphonate 98%, REacton®
Catalog # 43393.09
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
Density:1.7
 :591.45 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:34622-08-7
Formula:C₃F₉NdO₉S₃
MDL Number:MFCD00192343