Order Entry
Italy
ContactUsLinkComponent
Bis(triphenylphosphine)nickel(II) bromide ≥99%
Health
Irritant
Bis(triphenylphosphine)nickel(II) bromide ≥99%
Catalog # 42192.06
CAS Number:  
Health
Irritant
Bis(triphenylphosphine)nickel(II) bromide ≥99%
Catalog # 42192.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :743.08 g/mol
Melting Pt:219…223 °C
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:14126-37-5
Formula:C₃₆H₃₀Br₂NiP₂
MDL Number:MFCD00015865