
Bismuth(III) trifluoromethanesulphonate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:88189-03-1
:
Formula:C₃BiF₉O₉S₃
MDL Number:MFCD02093669
:656.19 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Titration Complexometric:31.0 to 33.0 % Bi
Specification Test Results
Frequently Bought Together