Order Entry
Italy
ContactUsLinkComponent
Brij® L23-SO-(AP) 30% (v/v) in water
Irritant
Brij® L23-SO-(AP) 30% (v/v) in water
Certificates
undefined
Irritant
Brij® L23-SO-(AP) 30% (v/v) in water
Certificates

Specification Test Results

  • Appearance:
    Clear colorless Jelly substance
  • Assay:
    29 to 31 %

Specification Test Results

Related Information

Frequently Bought Together