
Brij® 30
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9002-92-0
:
Formula:C₂₀H₄₂O₅
MDL Number:MFCD00043063
:362.55 g/mol
Melting Pt:14 °C
Flash Pt:>149 °C
EINECS:500-002-6
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to light yellow Liquid or solid
- Infrared spectrum:Conforms
- Refractive index:1.4510 to 1.4530 (20°C, 589 nm)
- Water:≤1 %
- Acid value:≤1 mg KOH/g
- Hydroxyl value:145 to 165 mg KOH/g
Specification Test Results
Frequently Bought Together