Order Entry
Italy
ContactUsLinkComponent
1,1-Cyclobutanedicarboxylic acid for synthesis, Sigma-Aldrich®
Corrosive
1,1-Cyclobutanedicarboxylic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41260.0005
Supplier:  Merck
CAS Number:  
Corrosive
1,1-Cyclobutanedicarboxylic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41260.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Melting Point:
    156 - 158 °C
  • Molar Mass:
    144.12 g/mol

Specification Test Results

About this item

 :
 :144.13 g/mol
Boiling Pt:663 °C (1013 hPa)
Melting Pt:156…158 °C
Storage Temperature:Ambient
ADR:8,II
 :M-Clarity™ quality level = MQ200
EINECS:226-651-2
UN:3261
 :GHS05
CAS Number:5445-51-2
Formula:C₆H₈O₄
MDL Number:MFCD00001325