
1,3-Cyclohexanedione ≥97% may cont. up to 1% NaCl
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:504-02-9
:
Formula:C₆H₈O₂
MDL Number:MFCD00001585
:112.13 g/mol
Merck Index:14,3178
Boiling Pt:235 °C (1013 hPa)
Melting Pt:101…105 °C
Density:1.100
EINECS:207-980-0
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together