Order Entry
Italy
Orders LinkContactUsLinkComponent
1,3-Cyclopentanedione 99%

1,3-Cyclopentanedione 99%

1,3-Cyclopentanedione 99%
Nome del fornitore:  Thermo Fisher Scientific
Certificates
1,3-Cyclopentanedione 99%

2 Options Available Below

Product Details & Documents
Synonyms:  Cyclopentane-1,3-dione
  • CAS Number:3859-41-4
  • Formula:C₅H₆O₂
  • MDL Number:MFCD00001405
  • Molecular Weight:98.1 g/mol
  • Melting Pt:148…153 °C
  • Storage Temperature:Refrigerator
  • EINECS:223-372-8

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Ochre to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    148 to 153 °C
  • Titration with NaOH:
    ≥98.5 %
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.