Order Entry
Italy
Orders LinkContactUsLinkComponent
Cyclohexanone, EMPLURA®, extra pure, Supelco®

Cyclohexanone, EMPLURA®, extra pure, Supelco®

Flammable
Corrosive
Irritant
   Sustainable Options Available
Supplier:   Merck
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Cyclohexanone, EMPLURA®, extra pure, Supelco®

5 Options Available Below

Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • UN:1915
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • REACH:01-2119453616-35
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1.02888.1000
  • 1.02888.2500
  • 1.02888.6010
  • 1.02888.6025
  • 1.02888.9191
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 113,00 €
    113,00 €
    1 * 1 L
       113,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    1.02888.1000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 236,00 €
    236,00 €
    1 * 2,5 L
       236,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    1.02888.2500
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
  • Stainless steel drum, returnable
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    1.02888.6010
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health
  • Stainless steel drum, returnable
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 1 370,00 €
    1 370,00 €
    1 * 25 L
       1 370,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    1.02888.6025
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health
  • Steel barrel
    190 L
    Log in for availability
    Log in for availability
    1 * 190 L
    1 * 190 L

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    1.02888.9191
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health