Order Entry
Italy
ContactUsLinkComponent
Cyclohexanone
Flammable
Irritant
Cyclohexanone
Catalog # EHERC11825500
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Flammable
Irritant
Cyclohexanone
Catalog # EHERC11825500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :98.14 g/mol
Merck Index:13,02755
Boiling Pt:155.6 °C (1013 hPa)
Melting Pt:–31 °C
Density:0.9478 g/cm³ (20 °C)
Flash Pt:43 °C
Storage Temperature:Ambient
EINECS:203-631-1
UN:1915
ADR:3,III
REACH:01-2119453616-35
 :GHS02
 :GHS07
CAS Number:108-94-1
Formula:C₆H₁₀(=O)
MDL Number:MFCD00001625