Order Entry
Italy
ContactUsLinkComponent
Cycloheptanone 98+%
Irritant
Flammable
Cycloheptanone 98+%
Certificates
undefined
Irritant
Flammable
Cycloheptanone 98+%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4595 to 1.4635 (20°C, 589 nm)
  • GC:
    ≥98.0 %

Specification Test Results

Related Information

Frequently Bought Together