

Benzyl benzoate 99.0-100.5% Ph. Eur.
Catalog # 21858.368
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Assay:99.0 to 100.5 %
- Appearance:Clear colourless liquid
- Identification A:Passes test
- Acidity:Passes test
- Relative density:1.118 to 1.122
- Refractive index (20°C):1.568 to 1.570
- Solidification point:Min. 17.0 °C
- Sulphated ash:Max. 0.1 %
- Residual solvents:Passes test
Specification Test Results
About this item
:
:212.25 g/mol
Merck Index:13,01129
Boiling Pt:323 °C (1013 hPa)
Melting Pt:17…20 °C
Density:1.118 g/cm³ (20 °C)
Flash Pt:158 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-402-9
UN:3082
ADR:9,III
:GHS07
:GHS09
CAS Number:120-51-4
Formula:C₁₄H₁₂O₂
MDL Number:MFCD00003075