


Mesitylene
Catalog # EHERC17881000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
Merck Index:13,05934
Boiling Pt:165 °C (1013 hPa)
Melting Pt:–45 °C
Density:0.8637 g/cm³ (20 °C)
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:203-604-4
UN:2325
ADR:3,III
:120.19 g/mol
:GHS02
:GHS07
:GHS09
CAS Number:108-67-8
Formula:C₉H₁₂
MDL Number:MFCD00008538