


1,2,4-Trimetilbenzene per sintesi, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:95-63-6
:
Formula:C₉H₁₂
MDL Number:MFCD00008527
:120.19 g/mol
Merck Index:12,08100
Boiling Pt:169 °C (1013 hPa)
Melting Pt:–44 °C
Density:0,88 g/cm³ (20 °C)
EINECS:202-436-9
UN:1993
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:44 °C
:GHS02
:GHS07
:GHS09
Storage Temperature:Ambiente
Specification Test Results
- Solubility:0.057 g/l (20 °C)
- Melting Point:-44 °C
- Molar Mass:120.19 g/mol
- Boiling Point:168 °C (1013 hPa)
- Vapor Pressure:2.8 hPa (25 °C)
- Flash Point:48 °C
- Refractive Index:1.5047 (20 °C, 589 nm)
- Explosion Limit:0.8 - 7 %(V)
- Density:0.88 g/cm3 (20 °C)
- Ignition Point:485 °C
Specification Test Results
Related Information
Frequently Bought Together