Biohazard Bags
Supplier: RATIOLAB
Certificates
About this item
These disposable waste bags are made from strong PP film with a broad seam at the bottom. Intended for the sterilisation and disposal of hazardous and contaminated labware.
- Rectangle white indicator patch field turns into dark when autoclaved
- Bags must not be completely closed while autoclaving
Available with biohazard sticker and safety imprint in English, German, French and Spanish language.
Frequently Bought Together