Order Entry
Italy
Orders LinkContactUsLinkComponent
3,4-Dimethylanisole ≥99%

3,4-Dimethylanisole ≥99%

Glass bottle for solids, 50 g
Catalog # B22735.18
3,4-Dimethylanisole ≥99%
Catalog # B22735.18
CAS Number:  4685-47-6
3,4-Dimethylanisole ≥99%
3,4-Dimethylanisole ≥99%

Glass bottle for solids, 50 g

CAS Number:  4685-47-6
Price
71,90 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 99% CAS No.: 4685-47-6 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 75°(167°F) Refractive Index: 1.5175 Boiling Point: 200-201° Beilstein Reference: 2040907 MDL No.: MFCD00008396

  • CAS Number:4685-47-6
  • Formula:C₉H₁₂O
  • MDL Number:MFCD00008396
  • Molecular Weight:136.19 g/mol
  • Boiling Pt:200…201 °C
  • Density:0.975
  • Flash Pt:75 °C (167 °F)
  • Storage Temperature:Ambient
  • EINECS:225-142-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.