Order Entry
Italy
ContactUsLinkComponent
2,3-Dimethylaniline for synthesis, Sigma-Aldrich®
Toxic
Health
Environment
2,3-Dimethylaniline for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Health
Environment
2,3-Dimethylaniline for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1.5 g/l (20 °C)
  • Melting Point:
    2 °C
  • Molar Mass:
    121.18 g/mol
  • Boiling Point:
    224 °C
  • Vapor Pressure:
    0.13 hPa (25 °C)
  • Flash Point:
    96 °C
  • Refractive Index:
    1.5685 (20 °C, 589 nm)
  • Explosion Limit:
    1 - 2.7 %(V)
  • Density:
    0.99 g/cm3 (20 °C)
  • Ignition Point:
    545 °C

Specification Test Results

Related Information

Frequently Bought Together