2,3-Dimethylaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2,3-Xylidine, 3-Amino-o-xylene
CAS Number:87-59-2
:
Formula:C₈H₁₁N
MDL Number:MFCD00007732
Boiling Pt:221…222 °C
Melting Pt:2…3 °C
Density:0.992
Molecular Weight:121.18 g/mol
Flash Pt:96 °C (204 °F)
Storage Temperature:Ambient
UN:1711
ADR:6.1 II
Merck Index:12,10217
EINECS:201-755-0
:GHS06
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together