

2-Methyl-3-nitrobenzotrifluoride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6656-49-1
:
Formula:C₈H₆F₃NO₂
MDL Number:MFCD00042322
:205.14 g/mol
Boiling Pt:86 °C (10 mmHg)
Flash Pt:>100 °C (212 °F)
EINECS:229-688-2
UN:2810
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together