Fmoc-D-Pro-OH
Supplier: Novabiochem (Part of Merck)
Jump to:
About this item
Synonyms: N-Fmoc-D-proline
- CAS Number:101555-62-8
- Formula:C₂₀H₁₉NO₄
- MDL Number:MFCD00077067
- Molecular Weight:337.38 g/mol
2 Options Available Below
Product Details & Documents
Synonyms: N-Fmoc-D-proline
- CAS Number:101555-62-8
- Formula:C₂₀H₁₉NO₄
- MDL Number:MFCD00077067
- Molecular Weight:337.38 g/mol
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



