Order Entry
Italy
Orders LinkContactUsLinkComponent
Fmoc-Pro-OH

Fmoc-Pro-OH

Plastic bottle for solids, 25 g
Catalog # 04-12-1031-25
Fmoc-Pro-OH
Catalog # 04-12-1031-25
CAS Number:  71989-31-6
Fmoc-Pro-OH

Plastic bottle for solids, 25 g

CAS Number:  71989-31-6
Price
35,20 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline
  • CAS Number:71989-31-6
  • Formula:C₂₀H₁₉NO₄
  • MDL Number:MFCD00037122
  • Molecular Weight:337.38 g/mol
  • EINECS:276-259-0

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g

Specifications