

Lidocaine ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:137-58-6
:
Formula:C₁₄H₂₂N₂O
MDL Number:MFCD00026733
Merck Index:14,05482
Boiling Pt:182°C
:234.34 g/mol
EINECS:205-302-8
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:66.0 to 70.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together