Pesticide reference standard, 1-Fluoronaphthalene 2,000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Catalog # EHERYA13794000MB
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:321-38-0
- Description:1-Fluoronaphthalene 2,000 µg/ml in tert-butyl methyl ether
- Formula:C₁₀H₇F
- Description:1-Fluoronaphthalene