Order Entry
Italy
ContactUsLinkComponent
Pesticide reference standard, 2-Chloronaphthalene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Pesticide reference standard, 2-Chloronaphthalene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20425200AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 2-Chloronaphthalene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20425200AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    91-58-7
  • Description:
    2-Chloronaphthalene 10 µg/ml in acetonitrile
  • Formula:
    C₁₀H₇Cl
  • Description:
    2-Chloronaphthalene

Specifications

About this item