2,4-Dinitro-N-phenylaniline
Codice catalogo EHERC12783702
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:259.22 g/mol
EINECS:213-508-4
CAS Number:961-68-2
Formula:C₁₂H₉N₃O₄
MDL Number:MFCD00007223