
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23364-44-5
:
Formula:C₁₄H₁₅NO
MDL Number:MFCD00074959
:213.28 g/mol
Melting Pt:142…144 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:141 to 145 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together