3-(Methylthio)phenylboronic acid
Catalog # MAYBAC35970.250
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:168.02 g/mol
CAS Number:128312-11-8
Formula:C₇H₉BO₂S
MDL Number:MFCD01319105