Tetrabutylammonium fluoride 1 M in THF
:429-41-4
:
:C₁₆H₃₆FN
:MFCD00011747
:0.887 g/cm³
:–17 °C
:Refrigerator
:207-057-2
:2924
:3,II
:261.47 g/mol
:GHS07
:GHS05
:GHS02
:GHS08
- Appearance:Clear yellow to amber to light green to brown Solution
- Water:≤8 %
- Titration with HClO4:0.90 to 1.10 M
Frequently Bought Together