Order Entry
Italy
ContactUsLinkComponent
2,4-Dimethylbenzaldehyde for synthesis, Sigma-Aldrich®
2,4-Dimethylbenzaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
2,4-Dimethylbenzaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    -9 °C
  • Molar Mass:
    134.18 g/mol
  • Boiling Point:
    215 - 216 °C
  • Vapor Pressure:
    1 hPa (20 °C)
  • Flash Point:
    88 °C
  • Refractive Index:
    1.5492 (20 °C, 589 nm)
  • Density:
    1.02 g/cm3 (20 °C)
  • Hill Formula:
    C9H10O
  • Chemical Formula:
    2,4-(CH3)2C6H3CHO

Specification Test Results

Related Information

Frequently Bought Together