
4-Pentylphenol
Catalog # EHERC10246800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
Boiling Pt:251 °C (1013 hPa)
Melting Pt:23 °C
Density:0.9597 g/cm³ (20 °C)
Flash Pt:104 °C
Storage Temperature:Ambient
EINECS:239-015-4
UN:2430
ADR:8,III
:164.25 g/mol
:GHS05
CAS Number:14938-35-3
Formula:C₁₁H₁₆O
MDL Number:MFCD00020211