Order Entry
Italy
ContactUsLinkComponent
2,6-Dinitrotoluene
2,6-Dinitrotoluene
Catalog # EHERC12786400
Supplier:  EHRENSTORFER
CAS Number:  
2,6-Dinitrotoluene
Catalog # EHERC12786400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :182.14 g/mol
Boiling Pt:300 °C (1013 hPa)
Melting Pt:65.2 °C
Density:1.538 g/cm³ (15 °C)
Storage Temperature:Ambient
EINECS:210-106-0
UN:3454
ADR:6.1,II
 :GHS06
 :GHS08
CAS Number:606-20-2
Formula:C₇H₆N₂O₄
MDL Number:MFCD00007158