2,6-Dinitrotoluene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:606-20-2
:
Formula:C₇H₆N₂O₄
MDL Number:MFCD00007158
:182.14 g/mol
Boiling Pt:285 °C
Melting Pt:58…65 °C
Density:1.538 g/cm³ (15 °C)
Flash Pt:207 °C (404 °F)
Storage Temperature:Ambient
EINECS:210-106-0
UN:3454
ADR:6.1,II
:GHS06
:GHS08
Related Information
Frequently Bought Together