3-Chlorotoluene 97%
:108-41-8
:
:C₇H₇Cl
:MFCD00000595
:13,02190
:160…162 °C (760 mmHg)
:–48 °C
:1.07 g/cm³
:50 °C
:Ambient
:2238
:3,III
:126.59 g/mol
:203-580-5
:GHS07
:GHS09
:GHS02
- Environmentally Preferable:
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5210 to 1.5230 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together