Order Entry
Italy
ContactUsLinkComponent
2,4-Dichlorotoluene for synthesis, Sigma-Aldrich®
Environment
2,4-Dichlorotoluene for synthesis, Sigma-Aldrich®
Catalog # 8.20458.9030
Supplier:  Merck
CAS Number:  
Environment
2,4-Dichlorotoluene for synthesis, Sigma-Aldrich®
Catalog # 8.20458.9030
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    30 kg
  • Seq:
    4319

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    -13 °C
  • Molar Mass:
    161.03 g/mol
  • Boiling Point:
    199 - 201 °C (1013 hPa)
  • Vapor Pressure:
    4 hPa (50 °C)
  • Flash Point:
    79 °C
  • Refractive Index:
    1.5454 (20 °C, 589 nm)
  • Density:
    1.25 g/cm3 (20 °C)
  • Ignition Point:
    450 °C

Specification Test Results

About this item

 :
 :161.03 g/mol
Boiling Pt:200 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.2498 g/cm³ (20 °C)
Flash Pt:79 °C
Storage Temperature:Ambient
EINECS:202-445-8
UN:3082
ADR:9,III
 :M-Clarity™ quality level = MQ200
 :GHS09
CAS Number:95-73-8
Formula:C₇H₆Cl₂
MDL Number:MFCD00000583