3-Nitro-o-toluic acid
Catalog #
Product Details & Documents
Synonyms: 2-Methyl-3-nitrobenzoic acid
- CAS Number:1975-50-4
- Formula:C₈H₇NO₄
- MDL Number:MFCD00007160
- Molecular Weight:181.15 g/mol
- Melting Pt:181…184 °C
- Storage Temperature:Ambient
- EINECS:217-826-4



