Order Entry
Italy
ContactUsLinkComponent
4-tert-Butyl-1-bromobenzene 97%
Irritant
4-tert-Butyl-1-bromobenzene 97%
Certificates
undefined
Irritant
4-tert-Butyl-1-bromobenzene 97%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5315 to 1.5345 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together