Order Entry
Italy
ContactUsLinkComponent
α,4-Dibromotoluene 98%
Corrosive
α,4-Dibromotoluene 98%
Certificates
undefined
Corrosive
α,4-Dibromotoluene 98%

Specification Test Results

  • Appearance:
    White to orange to tan Powder or crystals or needles and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    60 to 64 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together