
6-Chloro-2-pyridineboronic acid pinacol ester tech. 90%, Technical Grade
Catalog # H27426.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:239.51 g/mol
Melting Pt:107…110 °C
:GHS07
Storage Temperature:Freezer
CAS Number:652148-92-0
Formula:C₁₁H₁₅BClNO₂
MDL Number:MFCD06798259