Order Entry
Italy
Orders LinkContactUsLinkComponent
2,6-Dibromotoluene ≥98%

2,6-Dibromotoluene ≥98%

2,6-Dibromotoluene ≥98%
Certificates
2,6-Dibromotoluene ≥98%
2,6-Dibromotoluene ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 69321-60-4 Molecular Formula: C7H6Br2 Formula Weight: 249.94 Refractive Index: 1.6060 Melting Point: 2-6° Boiling Point: 112-113°/7mm Beilstein Reference: 3235502 MDL No.: MFCD00013524

  • CAS Number:69321-60-4
  • Formula:C₇H₆Br₂
  • MDL Number:MFCD00013524
  • Molecular Weight:249.93 g/mol
  • Boiling Pt:112…113 °C (7 mmHg)
  • Melting Pt:2…6 °C
  • Density:1.812
  • Storage Temperature:Ambient
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.