Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Phenol, crystals Ph. Eur.

Toxic
Corrosive
Health
Supplier:   VWR Collection
Phenol, crystals Ph. Eur.
Supplier:  VWR Collection
Certificates
Phenol, crystals Ph. Eur.

1 Options Available Below

Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Boiling Pt:181.8 °C (1013 hPa)
  • Melting Pt:40.8 °C
  • Density:1.06 g/cm³ (20 °C)
  • Flash Pt:81 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:203-632-7
  • UN:1671
  • ADR:6.1,II
Product Family Options

Product Information