Order Entry
Italy
ContactUsLinkComponent
1-Naphthylamine hydrochloride, Sigma-Aldrich®
1-Naphthylamine hydrochloride, Sigma-Aldrich®
Catalog # 8.20866.0100
Supplier:  Merck
CAS Number:  
1-Naphthylamine hydrochloride, Sigma-Aldrich®
Catalog # 8.20866.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    37.7 g/l (20 °C)
  • Melting Point:
    272 - 275 °C
  • Molar Mass:
    179.65 g/mol

Specification Test Results

About this item

Synonyms: 1-Naphthylammonium chloride
 :
 :179.65 g/mol
EINECS:209-014-3
UN:2811
 :GHS07
Storage Temperature:Ambient
CAS Number:552-46-5
Formula:C₁₀H₁₀ClN
MDL Number:MFCD00036370
ADR:6.1 II
 :M-Clarity™ quality level = MQ200