Order Entry
Italy
ContactUsLinkComponent
 
 
 
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 98%, Sigma-Aldrich®
 
undefined
 
 
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 98%, Sigma-Aldrich®

 

  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Purity (HPLC):
    ≥98.0%
  • Solubility (Turbidity), c = 5 mg/ml (0.5%); Water:
    Clear
  • Solubility (Colour):
    Yellow to Very Dark Yellow and Yellow-Green
  • Recommended Retest Period:
    5 years

 

Frequently Bought Together