2-(Trimethylsilyl)thiazole
Catalog # 8.14407.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 ml
- Seq:2070
Specifications
About this item
:
:157.31 g/mol
CAS Number:79265-30-8
Formula:C₆H₁₁NSSi
MDL Number:MFCD00066274