Order Entry
Italy
ContactUsLinkComponent
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol 99%
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol 99%
Catalog # ACRO319630050
CAS Number:  
undefined
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol 99%
Catalog # ACRO319630050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    70 to 77 °C
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :199.04 g/mol
Melting Pt:70…77 °C
Storage Temperature:Ambient
EINECS:242-863-8
CAS Number:19184-65-7
Formula:C₅H₁₁BrO₃
MDL Number:MFCD00135306