Order Entry
Italy
ContactUsLinkComponent
Erythrosin B ≥95% (dye content), Sigma-Aldrich®
Erythrosin B ≥95% (dye content), Sigma-Aldrich®
Certificates
undefined
Erythrosin B ≥95% (dye content), Sigma-Aldrich®

Specification Test Results

  • Appearance (Color): Orange to Red-Orange:
    Conforms to Requirements
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Wave Length:
    526 - 534 nm
  • Extinction Coefficient:
    ≥102000
  • Extinction Coefficient C = 0.005g/l, 95% Methanol 5% 0.1M Na2CO3:
    ≥32000
  • Carbon:
    27.3 - 30.2 %
  • Solubility (Turbidity) 1mg/ml 1M NH4OH:
    Clear
  • Solubility (Color):
    Orange to Red

Specification Test Results

Frequently Bought Together