Diglycolic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-99-6
:
Formula:C₄H₆O₅
MDL Number:MFCD00004309
:134.09 g/mol
Melting Pt:140…145 °C
EINECS:203-823-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together