Order Entry
Italy
ContactUsLinkComponent
 
 
1-Octyl-3-methylimidazolium hexafluorophosphate 98+%
 
undefined
 
1-Octyl-3-methylimidazolium hexafluorophosphate 98+%

 

  • Appearance:
    Clear light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4230 to 1.4250 (20°C, 589 nm)
  • Water:
    ≤1.0 % (K.F.)
  • NMR (1H-NMR):
    ≥98.0 %

 

 

Frequently Bought Together