Order Entry
Italy
ContactUsLinkComponent
2,5-Diiodo-1-methylimidazole ≥98%
Irritant
2,5-Diiodo-1-methylimidazole ≥98%
Catalog # H51212.03
CAS Number:  
Irritant
2,5-Diiodo-1-methylimidazole ≥98%
Catalog # H51212.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:86026-81-5
 :
Formula:C₄H₄I₂N₂
MDL Number:MFCD02179540
 :333.9 g/mol
 :GHS07
Storage Temperature:Ambient