Bis(pentafluorophenyl)carbonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59483-84-0
:
Formula:C₁₃F₁₀O₃
MDL Number:MFCD00368353
:394.13 g/mol
Melting Pt:47…50 °C
Flash Pt:110 °C (230 °F)
Storage Temperature:Ambient
Related Information
Frequently Bought Together